Assay Details
| Gene Symbol: CERT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005599613.3 | XP_005599670.1 | 1317 | 88 | ||
| XM_005599614.3 | XP_005599671.1 | 1317 | 88 | |||
| XM_005599616.3 | XP_005599673.1 | 1315 | 88 | |||
| XM_005599617.3 | XP_005599674.1 | 1317 | 88 | |||
| XM_023618163.1 | XP_023473931.1 | 1315 | 88 | |||
More Information
Set Membership: |
|

CERT1
Genomic Map