Assay Details
| Gene Symbol: LOC100073305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504043.6 | XP_001504093.1 | 3707 | 94 | ||
| XM_023618242.1 | XP_023474010.1 | 3982 | 94 | |||
| XM_023618243.1 | XP_023474011.1 | 1425 | 94 | |||
| GenBank mRNA | JL638619.1 | - | 3 - 4 | 247 | 94 | |
More Information
Set Membership: |
|

LOC100073305
Genomic Map