Assay Details
| Gene Symbol: LOC100073305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504043.6 | XP_001504093.1 | 2601 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001504043.6 transcripts.
Set Membership: |
|

LOC100073305
Genomic Map