Assay Details
| Gene Symbol: LOC100073305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504043.6 | XP_001504093.1 | 3191 | 120 | ||
| XM_023618242.1 | XP_023474010.1 | 3466 | 120 | |||
| XM_023618243.1 | XP_023474011.1 | 909 | 120 | |||
More Information
Set Membership: |
|

LOC100073305
Genomic Map