Assay Details
| Gene Symbol: ZNF496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023618260.1 | XP_023474028.1 | 1589 | 88 | ||
| XM_023618261.1 | XP_023474029.1 | 1414 | 88 | |||
| XM_023618262.1 | XP_023474030.1 | 1548 | 88 | |||
| XM_023618263.1 | XP_023474031.1 | 1590 | 88 | |||
| XM_023618264.1 | XP_023474032.1 | 1062 | 88 | |||
| XM_023618265.1 | XP_023474033.1 | 1237 | 88 | |||
| XM_023618266.1 | XP_023474034.1 | 1073 | 88 | |||
| XM_023618267.1 | XP_023474035.1 | 1016 | 88 | |||
More Information
Set Membership: |
|

ZNF496
Genomic Map