Assay Details
| Gene Symbol: SULT1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493165.4 | XP_001493215.1 | 565 | 128 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SULT1C4| Gene Symbol: SULT1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493165.4 | XP_001493215.1 | 565 | 128 | ||
Set Membership: |
|