Assay Details
| Gene Symbol: SULT1C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501352.6 | XP_001501402.2 | 424 | 96 | ||
| XM_023618462.1 | XP_023474230.1 | 425 | 96 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SULT1C2| Gene Symbol: SULT1C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501352.6 | XP_001501402.2 | 424 | 96 | ||
| XM_023618462.1 | XP_023474230.1 | 425 | 96 | |||
Set Membership: |
|