Assay Details
| Gene Symbol: LOC100062565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494086.6 | XP_001494136.2 | 389 | 62 | ||
| XM_023618680.1 | XP_023474448.1 | 390 | 62 | |||
More Information
Set Membership: |
|

LOC100062565
Genomic Map