Assay Details
| Gene Symbol: LOC102147863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005600244.3 | XP_005600301.2 | 1089 | 139 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102147863| Gene Symbol: LOC102147863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005600244.3 | XP_005600301.2 | 1089 | 139 | ||
Set Membership: |
|