Assay Details
| Gene Symbol: GEMIN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500469.5 | XP_001500519.1 | 1035 | 90 | ||
| XM_005600042.3 | XP_005600099.1 | 1033 | 90 | |||
More Information
Set Membership: |
|

GEMIN6
Genomic Map