Assay Details
| Gene Symbol: LOC100056590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023619476.1 | XP_023475244.1 | 241 | 99 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100056590| Gene Symbol: LOC100056590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023619476.1 | XP_023475244.1 | 241 | 99 | ||