Assay Details
| Gene Symbol: LOC100056590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503390.4 | XP_001503440.1 | 1426 | 97 | ||
| XM_001503392.4 | XP_001503442.1 | 1243 | 97 | |||
| XM_023619474.1 | XP_023475242.1 | 1288 | 97 | |||
| XM_023619476.1 | XP_023475244.1 | 1105 | 97 | |||

LOC100056590
Genomic Map