Assay Details
| Gene Symbol: CYRIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005600202.3 | XP_005600259.1 | 1640 | 84 | ||
| XM_005600208.3 | XP_005600265.1 | 3828 | 84 | |||
| XM_014731299.2 | XP_014586785.1 | 733 | 84 | |||
| XM_023619492.1 | XP_023475260.1 | 744 | 84 | |||
| XM_023619493.1 | XP_023475261.1 | 1529 | 84 | |||
| XM_023619494.1 | XP_023475262.1 | 1668 | 84 | |||
| XM_023619496.1 | XP_023475264.1 | 1509 | 84 | |||
| XM_023619497.1 | XP_023475265.1 | 1264 | 84 | |||
| XM_023619498.1 | XP_023475266.1 | 1118 | 84 | |||
| XM_023619499.1 | XP_023475267.1 | 888 | 84 | |||
| XM_023619500.1 | XP_023475268.1 | 890 | 84 | |||
| XM_023619501.1 | XP_023475269.1 | 521 | 84 | |||
| GenBank mRNA | JL615922.1 | - | 5 - 6 | 521 | 84 | |
More Information
Set Membership: |
|

CYRIA
Genomic Map