Assay Details
| Gene Symbol: SLC66A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502263.5 | XP_001502313.3 | 437 | 85 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC66A3| Gene Symbol: SLC66A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502263.5 | XP_001502313.3 | 437 | 85 | ||