Assay Details
| Gene Symbol: LSM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003363065.3 | XP_003363113.1 | 152 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003363065.3 transcripts.
Set Membership: |
|

LSM3
Genomic Map