Assay Details
| Gene Symbol: TAFA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014731366.2 | XP_014586852.2 | 4444 | 69 | ||
| XM_023620101.1 | XP_023475869.1 | 2513 | 69 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAFA1| Gene Symbol: TAFA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014731366.2 | XP_014586852.2 | 4444 | 69 | ||
| XM_023620101.1 | XP_023475869.1 | 2513 | 69 | |||
Set Membership: |
|