Assay Details
| Gene Symbol: LOC100059872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023620293.1 | XP_023476061.1 | 440 | 108 | ||
| XM_023620294.1 | XP_023476062.1 | 441 | 108 | |||
More Information
Set Membership: |
|

LOC100059872
Genomic Map