Assay Details
| Gene Symbol: LOC100059872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023620293.1 | XP_023476061.1 | 755 | 90 | ||
| XM_023620294.1 | XP_023476062.1 | 756 | 90 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100059872| Gene Symbol: LOC100059872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023620293.1 | XP_023476061.1 | 755 | 90 | ||
| XM_023620294.1 | XP_023476062.1 | 756 | 90 | |||