Assay Details
| Gene Symbol: C16H3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494391.6 | XP_001494441.3 | 1504 | 62 | ||
| XM_014731549.2 | XP_014587035.1 | 1511 | 62 | |||
More Information
Set Membership: |
|

C16H3orf84
Genomic Map