Assay Details
| Gene Symbol: C16H3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494391.6 | XP_001494441.3 | 1571 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16H3orf84| Gene Symbol: C16H3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494391.6 | XP_001494441.3 | 1571 | 72 | ||
Set Membership: |
|