Assay Details
| Gene Symbol: LOC100064912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495668.5 | XP_001495718.4 | 1500 | 65 | ||
| XM_005600740.3 | XP_005600797.2 | 874 | 65 | |||
| XM_005600742.3 | XP_005600799.2 | 493 | 65 | |||
| XM_005600743.3 | XP_005600800.2 | 486 | 65 | |||
| XM_023620549.1 | XP_023476317.1 | 1395 | 65 | |||
| XM_023620550.1 | XP_023476318.1 | 531 | 65 | |||
More Information
Set Membership: |
|

LOC100064912
Genomic Map