Assay Details
| Gene Symbol: OXSM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005600895.3 | XP_005600952.1 | 4019 | 76 | ||
| XM_014731837.2 | XP_014587323.1 | 1191 | 76 | |||
| XM_023620833.1 | XP_023476601.1 | 1783 | 76 | |||
More Information
Set Membership: |
|

OXSM
Genomic Map