Assay Details
| Gene Symbol: LOC100061057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023620859.1 | XP_023476627.1 | 491 | 133 | ||
| XM_023620860.1 | XP_023476628.1 | 393 | 133 | |||
| XM_023620861.1 | XP_023476629.1 | 337 | 133 | |||
More Information
Set Membership: |
|

LOC100061057
Genomic Map