Assay Details
| Gene Symbol: SEPTIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023644074.1 | XP_023499842.1 | 800 | 82 | ||
| XM_023644075.1 | XP_023499843.1 | 954 | 82 | |||
| XM_023644076.1 | XP_023499844.1 | 741 | 82 | |||
| XM_023644077.1 | XP_023499845.1 | 882 | 82 | |||
More Information
Set Membership: |
|

SEPTIN2
Genomic Map