Assay Details
| Gene Symbol: ADA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023642869.1 | XP_023498637.1 | 650 | 63 | ||
| XM_023642871.1 | XP_023498639.1 | 650 | 63 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ADA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023642869.1 | XP_023498637.1 | 650 | 63 | ||
| XM_023642871.1 | XP_023498639.1 | 650 | 63 | |||
Set Membership: |
|