Assay Details
| Gene Symbol: WASHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610835.3 | XP_005610892.1 | 1041 | 125 | ||
| XM_005610836.3 | XP_005610893.1 | 922 | 125 | |||
| XM_023642977.1 | XP_023498745.1 | 1118 | 125 | |||
More Information
Set Membership: |
|

WASHC1
Genomic Map