Assay Details
| Gene Symbol: SPART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495257.4 | XP_001495307.1 | 1782 | 75 | ||
| XM_005601141.3 | XP_005601198.1 | 2341 | 75 | |||
| XM_005601143.3 | XP_005601200.1 | 1685 | 75 | |||
| XM_005601144.3 | XP_005601201.1 | 1855 | 75 | |||
| GenBank mRNA | JL638549.1 | - | 5 - 6 | 802 | 75 | |
More Information
Set Membership: |
|

SPART
Genomic Map