Assay Details
| Gene Symbol: KLF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014732129.2 | XP_014587615.1 | 957 | 96 | ||
| XM_023621686.1 | XP_023477454.1 | 1537 | 96 | |||
| XM_023621687.1 | XP_023477455.1 | 960 | 96 | |||
More Information
Set Membership: |
|

KLF5
Genomic Map