Assay Details
| Gene Symbol: MAP3K20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499196.5 | XP_001499246.5 | 650 | 90 | ||
| XM_005601584.3 | XP_005601641.1 | 825 | 90 | |||
| XM_023622007.1 | XP_023477775.1 | 358 | 90 | |||
| XM_023622008.1 | XP_023477776.1 | 812 | 90 | |||
| XM_023622009.1 | XP_023477777.1 | 509 | 90 | |||
More Information
Set Membership: |
|

MAP3K20
Genomic Map