Assay Details
| Gene Symbol: FLACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014732327.2 | XP_014587813.1 | 1619 | 88 | ||
| XM_014732330.2 | XP_014587816.1 | 1388 | 88 | |||
| XM_023622356.1 | XP_023478124.1 | 1707 | 88 | |||
| XM_023622357.1 | XP_023478125.1 | 1297 | 88 | |||
| XM_023622358.1 | XP_023478126.1 | 1708 | 88 | |||
| GenBank mRNA | JL640648.1 | - | 1 - 2 | 466 | 88 | |
More Information
Set Membership: |
|

FLACC1
Genomic Map