Assay Details
| Gene Symbol: NAALADL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495653.5 | XP_001495703.2 | 2089 | 89 | ||
| XM_014732760.2 | XP_014588246.1 | 2484 | 89 | |||
More Information
Set Membership: |
|

NAALADL2
Genomic Map