Assay Details
| Gene Symbol: ADIPOQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499514.5 | XP_001499564.1 | 116 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001499514.5 transcripts.
Set Membership: |
|

ADIPOQ
Genomic Map