Assay Details
| Gene Symbol: GMNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014732813.2 | XP_014588299.1 | 667 | 101 | ||
| XM_023623498.1 | XP_023479266.1 | 691 | 101 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GMNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014732813.2 | XP_014588299.1 | 667 | 101 | ||
| XM_023623498.1 | XP_023479266.1 | 691 | 101 | |||
Set Membership: |
|