Assay Details
| Gene Symbol: TM4SF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023623570.1 | XP_023479338.1 | 272 | 69 | ||
| XM_023623571.1 | XP_023479339.1 | 272 | 69 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TM4SF19| Gene Symbol: TM4SF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023623570.1 | XP_023479338.1 | 272 | 69 | ||
| XM_023623571.1 | XP_023479339.1 | 272 | 69 | |||
Set Membership: |
|