Assay Details
| Gene Symbol: CFAP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014732868.2 | XP_014588354.2 | 1982 | 101 | ||
| XM_023623702.1 | XP_023479470.1 | 2435 | 101 | |||
| XM_023623703.1 | XP_023479471.1 | 2435 | 101 | |||
| XM_023623704.1 | XP_023479472.1 | 2440 | 101 | |||
| XM_023623705.1 | XP_023479473.1 | 2439 | 101 | |||
| XM_023623706.1 | XP_023479474.1 | 2441 | 101 | |||
| XM_023623707.1 | XP_023479475.1 | 1617 | 101 | |||
More Information
Set Membership: |
|

CFAP91
Genomic Map