Assay Details
| Gene Symbol: LOC111769099 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023623788.1 | XP_023479556.1 | 1185 | 102 | ||
| XM_023623789.1 | XP_023479557.1 | 1073 | 102 | |||
More Information
Set Membership: |
|

LOC111769099
Genomic Map