Assay Details
| Gene Symbol: PLPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001488028.5 | XP_001488078.1 | 827 | 92 | ||
| XM_005607020.3 | XP_005607077.1 | 378 | 92 | |||
| XM_005607021.3 | XP_005607078.1 | 834 | 92 | |||
| XM_023629111.1 | XP_023484879.1 | 831 | 92 | |||
More Information
Set Membership: |
|

PLPP3
Genomic Map