Assay Details
| Gene Symbol: TUT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023636810.1 | XP_023492578.1 | 1863 | 148 | ||
| XM_023636812.1 | XP_023492580.1 | 1043 | 148 | |||
| XM_023636813.1 | XP_023492581.1 | 1043 | 148 | |||
| XM_023636814.1 | XP_023492582.1 | 1043 | 148 | |||
| XM_023636815.1 | XP_023492583.1 | 1043 | 148 | |||
| XM_023636816.1 | XP_023492584.1 | 1043 | 148 | |||
| XM_023636817.1 | XP_023492585.1 | 1043 | 148 | |||
More Information
Set Membership: |
|

TUT4
Genomic Map