Assay Details
| Gene Symbol: C2H1orf109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503585.4 | XP_001503635.1 | 361 | 76 | ||
| XM_023633229.1 | XP_023488997.1 | 361 | 76 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001503585.4, XM_023633229.1 transcripts.
Set Membership: |
|

C2H1orf109
Genomic Map