Assay Details
| Gene Symbol: YARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023634189.1 | XP_023489957.1 | 670 | 98 | ||
| XM_023634193.1 | XP_023489961.1 | 633 | 98 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

YARS1| Gene Symbol: YARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023634189.1 | XP_023489957.1 | 670 | 98 | ||
| XM_023634193.1 | XP_023489961.1 | 633 | 98 | |||
Set Membership: |
|