Assay Details
| Gene Symbol: ZBTB8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014737455.2 | XP_014592941.1 | 1115 | 98 | ||
| XM_023634279.1 | XP_023490047.1 | 2004 | 98 | |||
| XM_023634281.1 | XP_023490049.1 | 2067 | 98 | |||
| XM_023634285.1 | XP_023490053.1 | 1408 | 98 | |||
| XM_023634294.1 | XP_023490062.1 | 1762 | 98 | |||
| XM_023634300.1 | XP_023490068.1 | 1818 | 98 | |||
More Information
Set Membership: |
|

ZBTB8A
Genomic Map