Assay Details
| Gene Symbol: ATP5IF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503993.5 | XP_001504043.1 | 164 | 119 | ||
| XM_023635050.1 | XP_023490818.1 | 332 | 119 | |||
More Information
Set Membership: |
|

ATP5IF1
Genomic Map