Assay Details
| Gene Symbol: CRYBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014737445.2 | XP_014592931.2 | 4660 | 107 | ||
| XM_023635414.1 | XP_023491182.1 | 4660 | 107 | |||
| XM_023635415.1 | XP_023491183.1 | 4660 | 107 | |||
| XM_023635416.1 | XP_023491184.1 | 4660 | 107 | |||
| XM_023635417.1 | XP_023491185.1 | 4660 | 107 | |||
More Information
Set Membership: |
|

CRYBG2
Genomic Map