Assay Details
| Gene Symbol: AUNIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504117.4 | XP_001504167.2 | 193 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001504117.4 transcripts.
Set Membership: |
|

AUNIP
Genomic Map