Assay Details
| Gene Symbol: LOC100071364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501109.6 | XP_001501159.2 | 164 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001501109.6 transcripts.
Set Membership: |
|

LOC100071364
Genomic Map