Assay Details
| Gene Symbol: C1QA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504261.4 | XP_001504311.1 | 128 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001504261.4 transcripts.
Set Membership: |
|

C1QA
Genomic Map