Assay Details
| Gene Symbol: CD69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499388.4 | XP_001499438.1 | 280 | 110 | ||
| GenBank mRNA | JL641191.1 | - | 2 - 3 | 363 | 110 | |
More Information
Set Membership: |
|

CD69
Genomic Map