Assay Details
| Gene Symbol: ART4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497341.4 | XP_001497391.1 | 504 | 85 | ||
| XM_014740635.2 | XP_014596121.1 | 504 | 85 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ART4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497341.4 | XP_001497391.1 | 504 | 85 | ||
| XM_014740635.2 | XP_014596121.1 | 504 | 85 | |||
Set Membership: |
|