Assay Details
| Gene Symbol: LOC100050294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023635700.1 | XP_023491468.1 | 659 | 89 | ||
| XM_023635702.1 | XP_023491470.1 | 659 | 89 | |||
| XM_023635703.1 | XP_023491471.1 | 453 | 89 | |||
More Information
Set Membership: |
|

LOC100050294
Genomic Map