Assay Details
| Gene Symbol: SPSB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494636.5 | XP_001494686.1 | 1021 | 62 | ||
| XM_023635846.1 | XP_023491614.1 | 1031 | 62 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPSB1| Gene Symbol: SPSB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001494636.5 | XP_001494686.1 | 1021 | 62 | ||
| XM_023635846.1 | XP_023491614.1 | 1031 | 62 | |||
Set Membership: |
|